missing translation for 'onlineSavingsMsg'
Learn More

Aniline Hydrobromide 98.0+%, TCI America™

Catalog No. p-7118180
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A29851G 1 g
A29855G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A29851G Supplier TCI America Supplier No. A29851G
Only null left

Chemical Identifiers

CAS 542-11-0
Molecular Formula C6H8BrN
Molecular Weight (g/mol) 174.041
MDL Number MFCD00035465
InChI Key KBPWECBBZZNAIE-UHFFFAOYSA-N
Synonym Anilinium Bromide
PubChem CID 68330
IUPAC Name aniline;hydrobromide
SMILES C1=CC=C(C=C1)N.Br

Specifications

Melting Point 285°C
Color White-Yellow
Quantity 1 g
Formula Weight 174.04
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Aniline Hydrobromide