accessibility menu, dialog, popup

UserName

CAS RN 542-11-0

IUPAC Name: hydrogen aniline bromide
Synonyms: hydrogen aniline bromide
Molecular Weight (g/mol): 174.04
Molecular Formula: C6H8BrN
InChi Key: KBPWECBBZZNAIE-UHFFFAOYSA-N
SMILES: [H+].[Br-].NC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)