missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aniline, ≥99.5% (GC), MilliporeSigma™ Supelco™
Supplier: Merck Emd Millipore 517885MLF
Description
EnvironmentalSpecifications
| 1139°F | |
| 0.7 mmHg (25°C) | |
| 1.022 g/mL (at 25°C) | |
| Neat | |
| MFCD00007629 | |
| 184°C | |
| 5 mL | |
| ≥99.5% (GC) |
| 62-53-3 | |
| 3.22 (185°C, vs. air) | |
| 70°C | |
| C6H5 NH2 | |
| -6°C | |
| Analytical Standard | |
| 93.13 | |
| Analytical Standard |