Learn More
Applied Biosystems™ Anhydrous Acetonitrile
This case contains 4 x 30 mL bottles of anhydrous acetonitrile.This anhydrous acetonitrile (amidite diluent) comes in a convenient large volume for ABI 394 Nucleic Acid synthesizers, resulting in extended hands-free DNA synthesis. With these larger bottles attached your synthesizer can produce up to 40 average primer-length oligonucleotides with no bottle changes, reducing attended operation time by 50%.
$149.00 - $487.00
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
400060
Encompass
View Documents Promotion Details
Applied Biosystems™
400060
30 mL
Each for $149.00
Only null left
4340863
Encompass
View Documents Promotion Details
Applied Biosystems™
4340863
4 x 30 mL
Each for $487.00
Only null left
Description
Description
Our high-performance reagents begin with raw materials that must conform to strict specifications. Once the reagents are manufactured and packaged, we perform extensive testing to assure quality. Reagent lifetime on the instrument is two weeks for tetrazole, and six weeks for all other reagents.
  • Reagent concentrations are within our strict specifications, guaranteeing consistent quality.
  • Our performance specification of at least 98% stepwise coupling efficiency is assured, increasing oligonucleotide synthesis yields of full-length product.
  • Reagent kits are available for your convenience, and save you time and money.
Videos
SDS
Documents
Documents
For Research Use Only. Not for use in diagnostic procedures.