Learn More
Agilent BioTek Angled Tip Injector Head for Imaging Multi-Mode Reader
Description
- 21° Angled tips for dispensing toward sides of microplate well to prevent cell layer disruption
- Includes installation instructions
Specifications
Specifications
| Product Type | Angled Tip Injector Head |
| For Use With (Equipment) | Imaging Multi-Mode Reader |
| For Use With (Application) | Dispensing towards sides of microplate well to prevent cell layer disruption. |