missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Angiopoietin 1 Monoclonal Antibody (9#)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA565971
Description
Angiopoietin 1 Monoclonal Antibody for Western Blot
Angiopoietins are proteins with important roles in vascular development and angiogenesis. All angiopoietins bind with similar affinity to an endothelial cell-specific tyrosine-protein kinase receptor. The protein encoded by this gene is a secreted glycoprotein that activates the receptor by inducing its tyrosine phosphorylation. It plays a critical role in mediating reciprocal interactions between the endothelium and surrounding matrix and mesenchyme. The protein also contributes to blood vessel maturation and stability, and may be involved in early development of the heart.Specifications
| Angiopoietin 1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| O08538, Q15389 | |
| Angpt1 | |
| Recombinant Angiopoietin 1 (ANGPT1) (Arg284-Cys452). | |
| 200 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 κ |
| Western Blot | |
| 9# | |
| Unconjugated | |
| Angpt1 | |
| 1110046O21Rik; 2810039D03Rik; AGP1; Agpt; Agpt1; ANG1; ang-1; ANG3; ANG-3; angioarrestin; angiopoietin 1; angiopoietin 3; angiopoietin like 1; angiopoietin Y1; angiopoietin-1; Angiopoietin-3; angiopoietin-like 1; angiopoietin-like protein 1; Angiopoietin-related protein 1; Angpt1; ANGPT3; Angptl1; AngY; ARP1; dJ595C2.2; dJ595C2.2 (angiopoietin Y1); endothelial growth factor; KIAA0003; PSEC0154; tie2 receptor ligand; UNQ162; UNQ162/PRO188 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 11600, 284 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |