missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Angiogenin Monoclonal Antibody (26-2F), eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 14976282
Encompass
Change view
Click to view available options
Quantity:
100 μg
2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14976282 100 μg
14976237 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14976282 Supplier Invitrogen™ Supplier No. 14976282
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse Monoclonal Antibody

The MAb 26-2F recognizes human angiogenin. Angiogenin is a secreted ribonuclease (RNASE)-family member and is important for the formation of new blood vessels as well as the stimulation of cell proliferation and survival. Cellular stress signals direct angiogenin to stress granules where angiogenin functions by hydrolyzing cellular tRNA resulting in modulation of protein translation and promotion of cell survival. Cytoplasmic angiogenin can bind to actin, forming complexes that activate protease cascades which can degrade laminin and fibronectin allowing endothelial cells to penetrate the basement membrane and migrate. In cancer cells, growth signals direct angiogenin translocation to the nucleus where binding of DNA promotor regions stimulates rRNA transcription resulting in a downstream increase in protein synthesis driving cell growth and survival. Angiogenin is upregulated in multiple cancers including breast, prostate, cervical, colorectal, lymphoma, and melanoma. During development and in adulthood, angiogenin is highly expressed in the spinal cord and contributes to motor neuron survival. Angiogenin is important for neuronal survival and is depleted in neurodegenerative diseases such as amyotrophic lateral sclerosis (ALS) and Parkinson's disease. This 26-2F antibody is reported to have both in vivo and in vitro neutralizing activity to angiogenin.

Angiogenin (ANG or ANG I) is important for the process of neovascularization and formation of new blood vessels. ANG is similar to pancreatic ribonuclease A and functions as a tRNA-specific ribonuclease that abolishes protein synthesis by specifically hydrolyzing cellular tRNAs. It interacts with endothelial cell-surface actin and may cause changes in the cell cytoskeleton. ANG is thought to be involved in the development of solid tumors and its antagonists are capable of inhibiting tumor growth. Defects in ANG are the cause of susceptibility to amyotrophic lateral sclerosis type 9 (ALS9). Angiogenin is a genetic link between ALS and PD.

Specifications

Antigen Angiogenin
Applications ELISA, Functional Assay, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Neutralization, Immunocytochemistry
Classification Monoclonal
Clone 26-2F
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene ANG
Gene Accession No. P03950
Gene Alias AI385586; ALS9; ANG; Ang1; Angiogenin; angiogenin ribonuclease 1; angiogenin, ribonuclease A family, member 1; angiogenin, ribonuclease, RNase A family, 5; angiogenin-1; epididymis luminal protein 168; HEL168; MGC22466; MGC71966; RAA1; Ribonuclease 5; ribonuclease A A1; ribonuclease A family member 5; RNase 5; RNASE4; RNASE5; Rnase5a
Gene Symbols ANG
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less