missing translation for 'onlineSavingsMsg'
Learn More

Andrographolide 98.0+%, TCI America™

Catalog No. A24591G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24591G 1 g
A24595G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A24591G Supplier TCI America Supplier No. A24591G

Chemical Identifiers

CAS 5508-58-7
Molecular Formula C20H30O5
Molecular Weight (g/mol) 350.46
MDL Number MFCD07778082
InChI Key BOJKULTULYSRAS-ZJFCSBQFSA-N
Synonym andrographolide, andrographis, unii-410105jhgr, s,e-4-hydroxy-3-2-1r,4as,5r,6r,8as-6-hydroxy-5-hydroxymethyl-5,8a-dimethyl-2-methylenedecahydronaphthalen-1-yl ethylidene dihydrofuran-2 3h-one, 3-2-decahydro-6-hydroxy-5-hydroxymethyl-5,8a-dimethyl-2-methylenenaphthyl ethylidene dihydro-4-hydroxyfuran-2 3h-one, 3alpha,14,15,18-tetrahydroxy-5b,9bh,10a-labda-8 20 ,12-dien-16-oic acid gamma-lactone, 1r-1-alpha e s ,4abeta,5alpha,6alpha,8aalpha-3-2-decahydro-6-hydroxy-5-hydroxymethyl-5,8a-dimethyl-2-methylene-1-naphthalenyl ethylidene dihydro-4-hydroxy-2 3h-furanone, 3e,4s-3-2-1r,4as,5r,6r,8as-6-hydroxy-5-hydroxymethyl-5,8a-dimethyl-2-methylidene-3,4,4a,6,7,8-hexahydro-1h-naphthalen-1-yl ethylidene-4-hydroxyoxolan-2-one, 3e,4s-3-2-1r,4as,5r,6r,8as-6-hydroxy-5-hydroxymethyl-5,8a-dimethyl-2-methylidene-hexahydro-1h-naphthalen-1-yl ethylidene-4-hydroxyoxolan-2-one, 3e,4s-4-hydroxy-3-2-1r,4as,5r,6r,8as-6-hydroxy-5-hydroxymethyl-5,8a-dimethyl-2-methylidenedecahydronaphthalen-1-yl ethylidene dihydrofuran-2 3h-one
PubChem CID 5318517
ChEBI CHEBI:65408
IUPAC Name (3E)-3-{2-[(1R,8aS)-6-hydroxy-5-(hydroxymethyl)-5,8a-dimethyl-2-methylidene-decahydronaphthalen-1-yl]ethylidene}-4-hydroxyoxolan-2-one
SMILES CC1(CO)C(O)CC[C@@]2(C)[C@H](C\C=C3/C(O)COC3=O)C(=C)CCC12

Specifications

Melting Point 231°C
Color White-Yellow
Quantity 1 g
Formula Weight 350.46
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Andrographolide
RTECSNumber LU3490750
TSCA No