missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ancymidol 98.0+%, TCI America™
$91.33 - $294.52
Chemical Identifiers
| CAS | 12771-68-5 |
|---|---|
| Molecular Formula | C15H16N2O2 |
| Molecular Weight (g/mol) | 256.31 |
| MDL Number | MFCD00072501 |
| InChI Key | HUTDUHSNJYTCAR-UHFFFAOYNA-N |
| Synonym | alpha-Cyclopropyl-alpha-(4-methoxyphenyl)-5-pyrimidinemethanol |
| PubChem CID | 25572 |
| ChEBI | CHEBI:73171 |
| IUPAC Name | cyclopropyl(4-methoxyphenyl)(pyrimidin-5-yl)methanol |
| SMILES | COC1=CC=C(C=C1)C(O)(C1CC1)C1=CN=CN=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
A287725MG
|
TCI America
A287725MG |
25 mg |
Each for $91.33
|
|
|||||
|
A2877100MG
|
TCI America
A2877100MG |
100 mg |
Each for $294.52
|
|
|||||
Chemical Identifiers
| 12771-68-5 | |
| 256.31 | |
| HUTDUHSNJYTCAR-UHFFFAOYNA-N | |
| 25572 | |
| cyclopropyl(4-methoxyphenyl)(pyrimidin-5-yl)methanol |
| C15H16N2O2 | |
| MFCD00072501 | |
| alpha-Cyclopropyl-alpha-(4-methoxyphenyl)-5-pyrimidinemethanol | |
| CHEBI:73171 | |
| COC1=CC=C(C=C1)C(O)(C1CC1)C1=CN=CN=C1 |
Specifications
| 12771-68-5 | |
| White-Yellow | |
| MFCD00072501 | |
| alpha-Cyclopropyl-alpha-(4-methoxyphenyl)-5-pyrimidinemethanol | |
| COC1=CC=C(C=C1)C(O)(C1CC1)C1=CN=CN=C1 | |
| 256.31 | |
| CHEBI:73171 | |
| ≥98.0% (GC) | |
| Ancymidol |
| 111°C | |
| C15H16N2O2 | |
| 25 mg | |
| HUTDUHSNJYTCAR-UHFFFAOYNA-N | |
| cyclopropyl(4-methoxyphenyl)(pyrimidin-5-yl)methanol | |
| 25572 | |
| 256.31 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : UV9280000
TSCA : No
Recommended Storage : Refrigerator