accessibility menu, dialog, popup

UserName

CAS RN 12771-68-5

CH3OOHNN

CAS RN 12771-68-5

IUPAC Name: cyclopropyl(4-methoxyphenyl)(pyrimidin-5-yl)methanol
Synonyms: ancymidol
Molecular Weight (g/mol): 256.31
Molecular Formula: C15H16N2O2
InChi Key: HUTDUHSNJYTCAR-UHFFFAOYNA-N
SMILES: COC1=CC=C(C=C1)C(O)(C1CC1)C1=CN=CN=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results