missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ Analytical Individual Weights: ASTM™ Class 1

Catalog No. 01912175
Change view
Click to view available options
Mass:
1 mg
2 mg
5 mg
10 mg
20 mg
50 mg
100 mg
200 mg
500 mg
1 g
2 g
5 g
10 g
20 g
50 g
100 g
200 g
500 g
1 kg
2 kg
5 kg
Product Type:
Analytical Sheet Weight
Two-Piece Analytical Weight
Includes:
Plastic box
Plastic box, No certificate
Plastic box; With certificate
Plastic box; Without certificate
32 product options available for selection
Product selection table with 32 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mass Product Type Includes
01912175 100 mg Analytical Sheet Weight Plastic box; With certificate
01912148 1 mg Analytical Sheet Weight Plastic box; Without certificate
01912169 1 mg Analytical Sheet Weight Plastic box; With certificate
01912170 2 mg Analytical Sheet Weight Plastic box; With certificate
01912150 5 mg Analytical Sheet Weight Plastic box; Without certificate
01912171 5 mg Analytical Sheet Weight Plastic box; With certificate
01912151 10 mg Analytical Sheet Weight Plastic box; Without certificate
01912172 10 mg Analytical Sheet Weight Plastic box; With certificate
01912173 20 mg Analytical Sheet Weight Plastic box; With certificate
01912174 50 mg Analytical Sheet Weight Plastic box; With certificate
01912154 100 mg Analytical Sheet Weight Plastic box; Without certificate
01912176 200 mg Analytical Sheet Weight Plastic box; With certificate
01912177 500 mg Analytical Sheet Weight Plastic box; With certificate
01912157 1 g Two-Piece Analytical Weight Plastic box; Without certificate
01912178 1 g Two-Piece Analytical Weight Plastic box; With certificate
01912179 2 g Two-Piece Analytical Weight Plastic box; With certificate
01912159 5 g Two-Piece Analytical Weight Plastic box
01912180 5 g Two-Piece Analytical Weight Plastic box; With certificate
01912181 10 g Two-Piece Analytical Weight Plastic box; With certificate
01912161 20 g Two-Piece Analytical Weight Plastic box; Without certificate
01912182 20 g Two-Piece Analytical Weight Plastic box; With certificate
01912183 50 g Two-Piece Analytical Weight Plastic box; With certificate
01912163 100 g Two-Piece Analytical Weight Plastic box, No certificate
01912184 100 g Two-Piece Analytical Weight Plastic box; With certificate
01912185 200 g Two-Piece Analytical Weight Plastic box; With certificate
01912165 500 g Two-Piece Analytical Weight Plastic box; Without certificate
01912186 500 g Two-Piece Analytical Weight Plastic box; With certificate
01912166 1 kg Two-Piece Analytical Weight Plastic box; Without certificate
01912187 1 kg Two-Piece Analytical Weight Plastic box; With certificate
01912167 2 kg Two-Piece Analytical Weight Plastic box
01912188 2 kg Two-Piece Analytical Weight Plastic box; With certificate
01912189 5 kg Two-Piece Analytical Weight Plastic box; With certificate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 32 options available.
32 options
Catalog No. 01912175 Supplier Mettler Toledo™ Supplier No. 11123475

Low magnetic permeability and high corrosion resistance for reliable stable values

Low magnetic permeability and high corrosion resistance for reliable stable values

  • ASTM Class 1
  • Premium stainless-steel, corrosion-resistant
  • Two-piece screw-knob construction

Specifications

Product Type Analytical Sheet Weight
Certifications/Compliance With certificate, ISO17025:2005, ISO9001, ISO14001 environmental standard, A2LA accredited lab Worthington, OH
Classification ASTM Class 1
Material Stainless Steel
Weight (Metric) 100 mg
Density 7.9kg/dm3 at 20°C
Mass 100 mg
Shape Square
Includes Plastic box; With certificate
Quantity Each
Packaging Quantity 1
Show More Show Less