missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Amylamine | |
| Yellow | |
| C5H13N | |
| 25 mL | |
| amylamine, 1-aminopentane, pentylamine, 1-pentylamine, n-amylamine, 1-pentanamine, n-pentylamine, monoamylamine, norleucamine, amyl amine | |
| CCCCCN | |
| 87.166 | |
| CHEBI:74848 | |
| ≥98.0% (GC) |
| 110-58-7 | |
| 104°C | |
| MFCD00008236 | |
| 1106 | |
| DPBLXKKOBLCELK-UHFFFAOYSA-N | |
| pentan-1-amine | |
| 8060 | |
| 87.17 | |
| Liquid |
Chemical Identifiers
| 110-58-7 | |
| 87.166 | |
| DPBLXKKOBLCELK-UHFFFAOYSA-N | |
| 8060 | |
| pentan-1-amine |
| C5H13N | |
| MFCD00008236 | |
| amylamine, 1-aminopentane, pentylamine, 1-pentylamine, n-amylamine, 1-pentanamine, n-pentylamine, monoamylamine, norleucamine, amyl amine | |
| CHEBI:74848 | |
| CCCCCN |
Safety and Handling
EINECSNumber : (2)-0133
RTECSNumber : SC0300000
TSCA : Yes