missing translation for 'onlineSavingsMsg'
Learn More

Amylamine 98.0+%, TCI America™

Catalog No. p-7116907
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A044525ML 25 mL
A0445100ML 100 mL
A0445500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A044525ML Supplier TCI America Supplier No. A044525ML
Only null left

Chemical Identifiers

CAS 110-58-7
Molecular Formula C5H13N
Molecular Weight (g/mol) 87.166
MDL Number MFCD00008236
InChI Key DPBLXKKOBLCELK-UHFFFAOYSA-N
Synonym amylamine, 1-aminopentane, pentylamine, 1-pentylamine, n-amylamine, 1-pentanamine, n-pentylamine, monoamylamine, norleucamine, amyl amine
PubChem CID 8060
ChEBI CHEBI:74848
IUPAC Name pentan-1-amine
SMILES CCCCCN

Specifications

Color Yellow
Boiling Point 104°C
Quantity 25 mL
UN Number 1106
Formula Weight 87.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Amylamine
EINECSNumber (2)-0133
RTECSNumber SC0300000
TSCA Yes