missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amyl salicylate, ≥98.0% (GC), MilliporeSigma™ Supelco™
Description
Cleaning Products, Cosmetics, Food and Beverages and Personal Care
Specifications
Specifications
| CAS | 2050-08-0 |
| Linear Formula | C12H16O3 |
| MDL Number | MFCD00082813 |
| Synonym | 2-Hydroxybenzoic acid pentyl ester; Pentyl salicylate; Salicylic acid amyl ester; Salicylic acid pentyl ester |
| Type | Analytical Standard |
| Quantity | 250 mg |
| Formula Weight | 208.25 |
| Percent Purity | ≥98.0% (GC) |
| Grade | Analytical Standard |