missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amyl Nitrite 95.0+%, TCI America™
Supplier: TCI America A0455100ML
Specifications
| Amyl Nitrite | |
| Yellow | |
| C5H11NO2 | |
| 100 mL | |
| Pentyl Nitrite | |
| CCCCCON=O | |
| 117.148 | |
| CHEBI:55344 | |
| ≥95.0% (GC) |
| 463-04-7 | |
| 104°C | |
| MFCD00036132 | |
| 1113 | |
| CSDTZUBPSYWZDX-UHFFFAOYSA-N | |
| pentyl nitrite | |
| 10026 | |
| 117.15 | |
| Liquid |
Chemical Identifiers
| 463-04-7 | |
| 117.148 | |
| CSDTZUBPSYWZDX-UHFFFAOYSA-N | |
| 10026 | |
| pentyl nitrite |
| C5H11NO2 | |
| MFCD00036132 | |
| Pentyl Nitrite | |
| CHEBI:55344 | |
| CCCCCON=O |
Safety and Handling
RTECSNumber : RA1140000
TSCA : Yes
Recommended Storage : Refrigerator