missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amyl n-Octanoate 98.0+%, TCI America™
$54.06 - $361.62
Chemical Identifiers
| CAS | 638-25-5 |
|---|---|
| Molecular Formula | C13H26O2 |
| Molecular Weight (g/mol) | 214.349 |
| MDL Number | MFCD00048919 |
| InChI Key | GJWGZSBNFSBUPX-UHFFFAOYSA-N |
| Synonym | amyl octanoate, amyl caprylate, octanoic acid, pentyl ester, amyl octoate, amyl octylate, pentyl octylate, amyl n-octanoate, n-caprylic acid n-amyl ester, n-amyl n-octanoate, unii-1kwp15429r |
| PubChem CID | 61185 |
| IUPAC Name | pentyl octanoate |
| SMILES | CCCCCCCC(=O)OCCCCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
O002825ML
|
TCI America
O002825ML |
25 mL |
Each for $54.06
|
|
|||||
|
O0028500ML
|
TCI America
O0028500ML |
500 mL |
Each for $361.62
|
|
|||||
Chemical Identifiers
| 638-25-5 | |
| 214.349 | |
| GJWGZSBNFSBUPX-UHFFFAOYSA-N | |
| 61185 | |
| CCCCCCCC(=O)OCCCCC |
| C13H26O2 | |
| MFCD00048919 | |
| amyl octanoate, amyl caprylate, octanoic acid, pentyl ester, amyl octoate, amyl octylate, pentyl octylate, amyl n-octanoate, n-caprylic acid n-amyl ester, n-amyl n-octanoate, unii-1kwp15429r | |
| pentyl octanoate |
Specifications
| 638-25-5 | |
| 260°C | |
| MFCD00048919 | |
| amyl octanoate, amyl caprylate, octanoic acid, pentyl ester, amyl octoate, amyl octylate, pentyl octylate, amyl n-octanoate, n-caprylic acid n-amyl ester, n-amyl n-octanoate, unii-1kwp15429r | |
| CCCCCCCC(=O)OCCCCC | |
| 214.349 | |
| 214.35 | |
| Liquid |
| Yellow | |
| C13H26O2 | |
| 25 mL | |
| GJWGZSBNFSBUPX-UHFFFAOYSA-N | |
| pentyl octanoate | |
| 61185 | |
| ≥98.0% (GC) | |
| Amyl n-Octanoate |
Safety and Handling
EINECSNumber : (2)-0782
TSCA : Yes