accessibility menu, dialog, popup

UserName

Explore New Spotlight Deals on Equipment, Instruments, and More

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 638-25-5

H3COOCH3

CAS RN 638-25-5

IUPAC Name: pentyl octanoate
Synonyms: amyl octanoate
Molecular Weight (g/mol): 214.35
Molecular Formula: C13H26O2
InChi Key: GJWGZSBNFSBUPX-UHFFFAOYSA-N
SMILES: CCCCCCCC(=O)OCCCCC
Boiling Point 
  • (2)
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)
1x1 image pixel for data collection 1x1 image pixel for data collection

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.