missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amyl Methyl Sulfide 97.0+%, TCI America™
$637.51 - $637.51
Chemical Identifiers
| CAS | 1741-83-9 |
|---|---|
| Molecular Formula | C6H14S |
| Molecular Weight (g/mol) | 118.24 |
| MDL Number | MFCD00027274 |
| InChI Key | FOJGPFUFFHWGFQ-UHFFFAOYSA-N |
| PubChem CID | 15620 |
| IUPAC Name | 1-(methylsulfanyl)pentane |
| SMILES | CCCCCSC |
Chemical Identifiers
| 1741-83-9 | |
| 118.24 | |
| FOJGPFUFFHWGFQ-UHFFFAOYSA-N | |
| 1-(methylsulfanyl)pentane |
| C6H14S | |
| MFCD00027274 | |
| 15620 | |
| CCCCCSC |
Specifications
| 1741-83-9 | |
| Yellow | |
| C6H14S | |
| 25 mL | |
| FOJGPFUFFHWGFQ-UHFFFAOYSA-N | |
| 1-(methylsulfanyl)pentane | |
| 15620 | |
| ≥97.0% (GC) | |
| Amyl Methyl Sulfide |
| -94°C | |
| 145°C | |
| MFCD00027274 | |
| 1993 | |
| CCCCCSC | |
| 118.24 | |
| 118.24 | |
| Liquid |
Safety and Handling
TSCA : No