missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amyl Laurate 97.0+%, TCI America™
$117.43 - $1,185.54
Chemical Identifiers
| CAS | 5350-03-8 |
|---|---|
| Molecular Formula | C17H34O2 |
| Molecular Weight (g/mol) | 270.46 |
| MDL Number | MFCD00059218 |
| InChI Key | PQWBDPUBNMEITD-UHFFFAOYSA-N |
| Synonym | Lauric Acid Amyl Ester, Pentyl Laurate, Lauric Acid Pentyl Ester |
| PubChem CID | 62571 |
| IUPAC Name | pentyl dodecanoate |
| SMILES | CCCCCCCCCCCC(=O)OCCCCC |
Chemical Identifiers
| 5350-03-8 | |
| 270.46 | |
| PQWBDPUBNMEITD-UHFFFAOYSA-N | |
| 62571 | |
| CCCCCCCCCCCC(=O)OCCCCC |
| C17H34O2 | |
| MFCD00059218 | |
| Lauric Acid Amyl Ester, Pentyl Laurate, Lauric Acid Pentyl Ester | |
| pentyl dodecanoate |
Specifications
| 5350-03-8 | |
| C17H34O2 | |
| 25 mL | |
| PQWBDPUBNMEITD-UHFFFAOYSA-N | |
| pentyl dodecanoate | |
| 62571 | |
| ≥97.0% (GC) | |
| Amyl Laurate |
| Yellow | |
| MFCD00059218 | |
| Lauric Acid Amyl Ester, Pentyl Laurate, Lauric Acid Pentyl Ester | |
| CCCCCCCCCCCC(=O)OCCCCC | |
| 270.46 | |
| 270.46 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0798
TSCA : No