accessibility menu, dialog, popup

UserName

CAS RN 5350-03-8

H3COOCH3

CAS RN 5350-03-8

IUPAC Name: pentyl dodecanoate
Synonyms: pentyl dodecanoate
Molecular Weight (g/mol): 270.46
Molecular Formula: C17H34O2
InChi Key: PQWBDPUBNMEITD-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCC(=O)OCCCCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results