missing translation for 'onlineSavingsMsg'
Learn More

Amyl Hexanoate 98.0+%, TCI America™

Catalog No. p-7130345
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H010625ML 25 mL
H0106500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H010625ML Supplier TCI America Supplier No. H010625ML

Chemical Identifiers

CAS 540-07-8
Molecular Formula C11H22O2
Molecular Weight (g/mol) 186.295
MDL Number MFCD00027281
InChI Key WRFZKAGPPQGDDQ-UHFFFAOYSA-N
Synonym Hexanoic Acid Amyl Ester, Pentyl Hexanoate, Hexanoic Acid Pentyl Ester
PubChem CID 10886
IUPAC Name pentyl hexanoate
SMILES CCCCCC(=O)OCCCCC

Specifications

Color Yellow
Boiling Point 226°C
Quantity 25 mL
Formula Weight 186.30
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Amyl Hexanoate
EINECSNumber (2)-0782
RTECSNumber MO8421700
TSCA Yes