accessibility menu, dialog, popup

UserName

CAS RN 540-07-8

H3COOCH3

CAS RN 540-07-8

IUPAC Name: pentyl hexanoate
Synonyms: amyl hexanoate
Molecular Weight (g/mol): 186.3
Molecular Formula: C11H22O2
InChi Key: WRFZKAGPPQGDDQ-UHFFFAOYSA-N
SMILES: CCCCCOC(=O)CCCCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results