missing translation for 'onlineSavingsMsg'
Learn More

Amyl Formate 95.0+%, TCI America™

Catalog No. p-7129462
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F004925ML 25 mL
F0049500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F004925ML Supplier TCI America Supplier No. F004925ML
Only null left

Chemical Identifiers

CAS 638-49-3
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00040440
InChI Key DIQMPQMYFZXDAX-UHFFFAOYSA-N
Synonym Formic Acid Amyl Ester, Pentyl Formate
PubChem CID 12529
IUPAC Name pentyl formate
SMILES CCCCCOC=O

Specifications

Color Colorless
Boiling Point 130°C
Quantity 25 mL
UN Number 1109
Formula Weight 116.16
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Amyl Formate
EINECSNumber (2)-0684
RTECSNumber LQ9370000
TSCA Yes