accessibility menu, dialog, popup

UserName

CAS RN 638-49-3

H3COO

CAS RN 638-49-3

IUPAC Name: pentyl formate
Synonyms: amyl formate
Molecular Weight (g/mol): 116.16
Molecular Formula: C6H12O2
InChi Key: DIQMPQMYFZXDAX-UHFFFAOYSA-N
SMILES: CCCCCOC=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results