missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amyl Formate 95.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 130°C |
| Quantity | 500 mL |
| UN Number | 1109 |
| Formula Weight | 116.16 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Amyl Formate |
Safety and Handling
| EINECSNumber | (2)-0684 |
| RTECSNumber | LQ9370000 |
| TSCA | Yes |