missing translation for 'onlineSavingsMsg'
Learn More

Amyl Chloroformate 97.0+%, TCI America™

Catalog No. C058725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C058725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C058725G Supplier TCI America Supplier No. C058725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 638-41-5
Molecular Formula C6H11ClO2
Molecular Weight (g/mol) 150.602
MDL Number MFCD00058933
InChI Key XHRRYUDVWPPWIP-UHFFFAOYSA-N
Synonym Chloroformic Acid Amyl Ester, Chloroformic Acid Pentyl Ester, Pentyl Chloroformate
PubChem CID 69489
IUPAC Name pentyl carbonochloridate
SMILES CCCCCOC(=O)Cl

Specifications

Color Colorless
Boiling Point 161°C
Quantity 25 g
UN Number 3277
Formula Weight 150.60
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Amyl Chloroformate
TSCA Yes
Recommended Storage Refrigerator