accessibility menu, dialog, popup

UserName

H3COClO

CAS RN 638-41-5

IUPAC Name: pentyl carbonochloridate
Synonyms: pentyl carbonochloridate
Molecular Weight (g/mol): 150.6
Molecular Formula: C6H11ClO2
InChi Key: XHRRYUDVWPPWIP-UHFFFAOYSA-N
SMILES: CCCCCOC(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results