missing translation for 'onlineSavingsMsg'
Learn More

Amyl Acetate, Technical, Spectrum™ Chemical

Catalog No. p-7165024
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
Packaging:
Amber Glass Bottle
Steel Can
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608861 500 mL Amber Glass Bottle
18608860 4 L Amber Glass Bottle
18608859 20 L Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18608861 Supplier Spectrum Chemical Mfg Cor Supplier No. A1283500MLGL
Only null left

Chemical Identifiers

CAS 628-63-7

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 500 mL
Grade Technical

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.