missing translation for 'onlineSavingsMsg'
Learn More

Amyl Acetate 99.0+%, TCI America™

Catalog No. A002125ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A002125ML 25 mL
A0021500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A002125ML Supplier TCI America Supplier No. A002125ML

Chemical Identifiers

CAS 628-63-7
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.187
MDL Number MFCD00009500
InChI Key PGMYKACGEOXYJE-UHFFFAOYSA-N
Synonym amyl acetate, n-amyl acetate, acetic acid, pentyl ester, n-pentyl acetate, amyl acetic ester, 1-pentyl acetate, n-pentyl ethanoate, birnenoel, pent-acetate, amyl acetic ether
PubChem CID 12348
ChEBI CHEBI:87362
IUPAC Name pentyl acetate
SMILES CCCCCOC(=O)C

Specifications

Color Colorless
Boiling Point 148°C
Quantity 25 mL
UN Number 1104
Formula Weight 130.19
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Amyl Acetate
EINECSNumber (2)-0733
RTECSNumber AJ1925000
TSCA Yes