missing translation for 'onlineSavingsMsg'
Learn More

Amyl Acetate, 98%, Spectrum™ Chemical

Catalog No. p-7165023
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608856 25 mL Amber Glass Bottle
18608853 100 mL Amber Glass Bottle
18608858 500 mL Amber Glass Bottle
18608857 4 L Amber Glass Bottle
18608854 20 L Steel Can
18608855 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18608856 Supplier Spectrum Chemical Mfg Cor Supplier No. A128025ML
Only null left

Chemical Identifiers

CAS 628-63-7
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.19
InChI Key PGMYKACGEOXYJE-UHFFFAOYSA-N
IUPAC Name pentyl acetate
SMILES CCCCCOC(C)=O

Specifications

Acidity 0.0001
CAS Max % 1
Packaging Amber Glass Bottle
Quantity 25 mL
Percent Purity 0.98
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.