missing translation for 'onlineSavingsMsg'
Learn More

Amyl Acetate, 98%, Spectrum™ Chemical

Catalog No. 18608855
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608855 200 L Drum
18608856 25 mL Amber Glass Bottle
18608853 100 mL Amber Glass Bottle
18608858 500 mL Amber Glass Bottle
18608857 4 L Amber Glass Bottle
18608854 20 L Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18608855 Supplier Spectrum Chemical Mfg Cor Supplier No. A1280200LTBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 628-63-7
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.19
InChI Key PGMYKACGEOXYJE-UHFFFAOYSA-N
IUPAC Name pentyl acetate
SMILES CCCCCOC(C)=O

Specifications

Acidity 0.0001
CAS Max % 1
Packaging Drum
Quantity 200 L
Percent Purity 0.98
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.