missing translation for 'onlineSavingsMsg'
Learn More

Amyl 4-Hydroxybenzoate 98.0+%, TCI America™

Catalog No. p-7130407
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H020810G 10 g
H020825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H020810G Supplier TCI America Supplier No. H020810G
Only null left

Chemical Identifiers

CAS 6521-29-5
Molecular Formula C12H16O3
Molecular Weight (g/mol) 208.26
MDL Number MFCD00016479
InChI Key ZNSSPLQZSUWFJT-UHFFFAOYSA-N
PubChem CID 23019
IUPAC Name pentyl 4-hydroxybenzoate
SMILES CCCCCOC(=O)C1=CC=C(O)C=C1

Specifications

Color White
Quantity 10 g
Synonym amyl 4-hydroxybenzoate, pentyl p-hydroxybenzoate, pentylparaben, n-pentyl 4-hydroxybenzoate, benzoic acid, 4-hydroxy-, pentyl ester, p-hydroxybenzoic acid pentyl ester, pentyl paraben, unii-sjv2e4c7dm, p-oxybenzoesaurepentylester, amyl4-hydroxybenzoate
Formula Weight 208.26
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material Amyl 4-Hydroxybenzoate
EINECSNumber (3)-1585
RTECSNumber DH2555000
TSCA No
Recommended Storage Refrigerator