missing translation for 'onlineSavingsMsg'
Learn More

Amsacrine Hydrochloride 98.0+%, TCI America™

Catalog No. p-7118066
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2777100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A2777100MG Supplier TCI America Supplier No. A2777100MG

Chemical Identifiers

CAS 54301-15-4
Molecular Formula C21H20ClN3O3S
Molecular Weight (g/mol) 429.919
MDL Number MFCD07799963
InChI Key WDISRLXRMMTXEV-UHFFFAOYSA-N
Synonym 4-(9-Acridinylamino)-N-(methanesulfonyl)-m-anisidine Hydrochloride, m-AMSA
PubChem CID 148673
IUPAC Name N-[4-(acridin-9-ylamino)-3-methoxyphenyl]methanesulfonamide;hydrochloride
SMILES COC1=C(C=CC(=C1)NS(=O)(=O)C)NC2=C3C=CC=CC3=NC4=CC=CC=C42.Cl

Specifications

Melting Point 288°C
Quantity 100 mg
UN Number 2811
Formula Weight 429.92
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Amsacrine Hydrochloride
RTECSNumber PB1081000
TSCA No