accessibility menu, dialog, popup

UserName

CAS RN 54301-15-4

IUPAC Name: hydrogen N-{4-[(acridin-9-yl)amino]-3-methoxyphenyl}methanesulfonamide chloride
Synonyms: hydrogen amsacrine chloride
Molecular Weight (g/mol): 429.92
Molecular Formula: C21H20ClN3O3S
InChi Key: WDISRLXRMMTXEV-UHFFFAOYSA-N
SMILES: [H+].[Cl-].COC1=CC(NS(C)(=O)=O)=CC=C1NC1=C2C=CC=CC2=NC2=CC=CC=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results