missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Amrinone, 98%

Phosphodiesterase III inhibitor and inhibits platelet aggregation | CAS: 60719-84-8 | C10H9N3O

Supplier:  Thermo Scientific Chemicals J6265503

Quantity 1 g

 View more versions of this product

Catalog No. AAJ6265503


Only null left

Description

Description

Phosphodiesterase III inhibitor and inhibits platelet aggregation

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

60719-84-8
C10H9N3O
5-Amino-(3,4′-bipyridin)-6(1H)-one

Specifications

1 g
2°C to 8°C
98%
C10H9N3O
5-Amino-(3,4′-bipyridin)-6(1H)-one
Insoluble in water or chloroform; Slightly soluble in acetic acid or DMF; Soluble in DMSO
Crystals
Yellow
Phosphodiesterase III inhibitor and inhibits platelet aggregation
60719-84-8
187.20g/mol
294°C to 297°C (dec.)
Signal Transduction Reagent
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 262-390-0

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions