missing translation for 'onlineSavingsMsg'
Learn More

Amprolium Hydrochloride 98.0+%, TCI America™

Catalog No. p-7117976
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A25725G 5 g
A257225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A25725G Supplier TCI America Supplier No. A25725G
Only null left

Chemical Identifiers

CAS 137-88-2
Molecular Formula C14H20Cl2N4
Molecular Weight (g/mol) 315.24
MDL Number MFCD00078831
InChI Key PJBQYZZKGNOKNJ-UHFFFAOYSA-M
Synonym amprolium hydrochloride, amprolium hcl, amprolium, mepyrium, amprol, amprol 25, thiacoccid, amprol 25 per cent, amprolsol vet, corid vet
PubChem CID 8732
IUPAC Name hydrogen 1-[(4-amino-2-propylpyrimidin-5-yl)methyl]-2-methylpyridin-1-ium dichloride
SMILES [H+].[Cl-].[Cl-].CCCC1=NC=C(C[N+]2=CC=CC=C2C)C(N)=N1

Specifications

Melting Point 241°C
Quantity 5 g
Formula Weight 315.24
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Amprolium Hydrochloride [for Biochemical Research]
RTECSNumber TJ7565000
TSCA No