missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amprolium Hydrochloride 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 241°C |
| Quantity | 5 g |
| Formula Weight | 315.24 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Amprolium Hydrochloride [for Biochemical Research] |
Safety and Handling
| RTECSNumber | TJ7565000 |
| TSCA | No |