missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amphotericin B, MP Biomedicals
$408.99 - $408.99
Chemical Identifiers
| CAS | 1397-89-3 |
|---|---|
| Molecular Formula | C47H73NO17 |
| Molecular Weight (g/mol) | 924.09 |
| MDL Number | MFCD00877763 |
| InChI Key | APKFDSVGJQXUKY-ZNVUZQDLSA-N |
| Synonym | Amphomoronal, Fungizone, Amphotericine B |
| PubChem CID | 134129663 |
| SMILES | C[C@H]1O[C@@H](O[C@@H]2C[C@@H]3O[C@@](O)(C[C@H](O)[C@H]3C(O)=O)C[C@@H](O)C[C@@H](O)[C@H](O)CC[C@@H](O)C[C@@H](O)CC(=O)O[C@@H](C)[C@H](C)[C@H](O)[C@@H](C)\C=C/C=C\C=C/C=C\C=C/C=C\C=C/2)[C@@H](O)[C@@H](N)[C@@H]1O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN19504390
|
MP Biomedicals Inc
0219504390 |
500 mg |
Each for $408.99
|
|
|||||
Chemical Identifiers
| 1397-89-3 | |
| 924.09 | |
| APKFDSVGJQXUKY-ZNVUZQDLSA-N | |
| 134129663 |
| C47H73NO17 | |
| MFCD00877763 | |
| Amphomoronal, Fungizone, Amphotericine B | |
| C[C@H]1O[C@@H](O[C@@H]2C[C@@H]3O[C@@](O)(C[C@H](O)[C@H]3C(O)=O)C[C@@H](O)C[C@@H](O)[C@H](O)CC[C@@H](O)C[C@@H](O)CC(=O)O[C@@H](C)[C@H](C)[C@H](O)[C@@H](C)\C=C/C=C\C=C/C=C\C=C/C=C\C=C/2)[C@@H](O)[C@@H](N)[C@@H]1O |
Specifications
| Amphotericin B | |
| 1397-89-3 | |
| 500 mg | |
| MFCD00877763 | |
| APKFDSVGJQXUKY-ZNVUZQDLSA-N | |
| (1S,3R,18S,19R,20R,21S,25R,27R,30R,31R,33S,35R,37S,38R)-3-[(2R,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-methyloxan-2-yl]oxy-19,25,27,30,31,33,35,37-octahydroxy-18,20,21-trimethyl-23-oxo-22,39-dioxabicyclo[33.3.1]nonatriaconta-4,6,8,10,12,14,16-heptaene-38-car | |
| 134129663 | |
| Amphotericin B, from Streptomyces sp. |
| Powder | |
| Yellow | |
| C47H73NO17 | |
| Amphomoronal, Fungizone, Amphotericine B | |
| C[C@H]1O[C@@H](O[C@@H]2C[C@@H]3O[C@@](O)(C[C@H](O)[C@H]3C(O)=O)C[C@@H](O)C[C@@H](O)[C@H](O)CC[C@@H](O)C[C@@H](O)CC(=O)O[C@@H](C)[C@H](C)[C@H](O)[C@@H](C)\C=C/C=C\C=C/C=C\C=C/C=C\C=C/2)[C@@H](O)[C@@H](N)[C@@H]1O | |
| 924.09 | |
| 924.1 |
Safety and Handling
EINECSNumber : 215-742-2