missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amoxicillin Trihydrate 98.0+%, TCI America™
$66.46 - $201.25
Chemical Identifiers
| CAS | 61336-70-7 |
|---|---|
| Molecular Formula | C16H25N3O8S |
| Molecular Weight (g/mol) | 419.449 |
| MDL Number | MFCD00072029 |
| InChI Key | MQXQVCLAUDMCEF-FXHJIJKUSA-N |
| PubChem CID | 124080955 |
| IUPAC Name | (2S,5R,6S)-6-[[(2S)-2-amino-2-(4-hydroxyphenyl)acetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;trihydrate |
| SMILES | CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=C(C=C3)O)N)C(=O)O)C.O.O.O |
Chemical Identifiers
| 61336-70-7 | |
| 419.449 | |
| MQXQVCLAUDMCEF-FXHJIJKUSA-N | |
| (2S,5R,6S)-6-[[(2S)-2-amino-2-(4-hydroxyphenyl)acetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;trihydrate |
| C16H25N3O8S | |
| MFCD00072029 | |
| 124080955 | |
| CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=C(C=C3)O)N)C(=O)O)C.O.O.O |
Specifications
| 61336-70-7 | |
| C16H25N3O8S | |
| White/Yellow | |
| CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=C(C=C3)O)N)C(=O)O)C.O.O.O | |
| 419.449 | |
| 124080955 | |
| ≥98.0% (T) |
| Powder/Crystals | |
| MFCD00072029 | |
| MQXQVCLAUDMCEF-FXHJIJKUSA-N | |
| (2S,5R,6S)-6-[[(2S)-2-amino-2-(4-hydroxyphenyl)acetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;trihydrate | |
| 5 g | |
| 365.40 | |
| Amoxicillin Trihydrate |
Safety and Handling
RTECSNumber : XH8300000
TSCA : No