missing translation for 'onlineSavingsMsg'
Learn More

Amoxicillin Trihydrate 98.0+%, TCI America™

Catalog No. A20995G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A20995G 5 g
A209925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A20995G Supplier TCI America Supplier No. A20995G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 61336-70-7
Molecular Formula C16H25N3O8S
Molecular Weight (g/mol) 419.449
MDL Number MFCD00072029
InChI Key MQXQVCLAUDMCEF-FXHJIJKUSA-N
PubChem CID 124080955
IUPAC Name (2S,5R,6S)-6-[[(2S)-2-amino-2-(4-hydroxyphenyl)acetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;trihydrate
SMILES CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=C(C=C3)O)N)C(=O)O)C.O.O.O

Specifications

Physical Form Powder/Crystals
Color White/Yellow
Quantity 5 g
Formula Weight 365.40
Percent Purity ≥98.0% (T)
Chemical Name or Material Amoxicillin Trihydrate
RTECSNumber XH8300000
TSCA No