missing translation for 'onlineSavingsMsg'
Learn More

Amoxicillin trihydrate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014889
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014889 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014889 Supplier Merck Emd Millipore Supplier No. PHR11271G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Amoxicillin trihydrate is a oral semisynthetic penicillin, which shows antibacterial activity against Gram positive and and a limited range of Gram-negative organisms. Its mode of action involves the inhibition of bacterial cell wall synthesis. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 61336-70-7
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C16H19N3O5 S · 3H2 O
MDL Number MFCD00072029
Type Certified Reference Material
Quantity 1 g
Formula Weight 419.45
Grade Certified Reference Material