missing translation for 'onlineSavingsMsg'
Learn More

Amoxapine Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012174
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012174 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012174 Supplier Merck Emd Millipore Supplier No. PHR1841500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Amoxapine is a tricyclic antidepressant known to inhibit reuptake of neuronal transmitter. The drug helps in controlling agitation and anxiety along with depression.Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 14028-44-5
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C17H16 ClN3 O
MDL Number MFCD00069210
Synonym Amoxapine; 2-Chloro-11-(1-piperazinyl)dibenz[b,f][1,4 ]oxazepine
Type Certified Reference Material
Quantity 500 mg
Formula Weight 313.78
Grade Certified Reference Material