missing translation for 'onlineSavingsMsg'
Learn More

Amorolfine hydrochloride, 98%

Catalog No. AC458752500
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC458752500 250 mg
AC458750010 1 g
AC458750050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC458752500 Supplier Thermo Scientific Chemicals Supplier No. 458752500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 78613-38-4 | C21H35NO·HCl | 353.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 78613-38-4
Molecular Formula C21H35NO·HCl
Molecular Weight (g/mol) 353.97
InChI Key XZKWIPVTHGWDCF-KUZYQSSXSA-N
Synonym amorolfine hydrochloride, amorolfine hcl, loceryl, curanail, locetar, odenil, amorolfin, pekiron, amorolfine hydrochloride jan
PubChem CID 54259
ChEBI CHEBI:59649
IUPAC Name (2R,6S)-2,6-dimethyl-4-[2-methyl-3-[4-(2-methylbutan-2-yl)phenyl]propyl]morpholine;hydrochloride
SMILES CCC(C)(C)C1=CC=C(C=C1)CC(C)CN2CC(OC(C2)C)C.Cl

Specifications

Melting Point 221.0°C to 223.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Quantity 250 mg
Solubility Information Solubility in water: 9g/L. Other solubilities: 28 mg/mL dmso, 71 mg/mL ethanol
Formula Weight 353.97
Percent Purity 98%
Chemical Name or Material Amorolfine hydrochloride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before re

RUO – Research Use Only