missing translation for 'onlineSavingsMsg'
Learn More

Amobarbital-D5 Solution, MilliporeSigma™ Supelco™

Catalog No. 01001000
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001000 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001000 Supplier Merck Emd Millipore Supplier No. A1021ML
Only null left

Certified reference material, Cerilliant™

Specifications

CAS 1190003-63-4
Molecular Weight (g/mol) 231.3
Quantity 1 mL