Learn More
Ammonium zinc sulfate hydrate
CAS: 77998-33-5 | H8N2O8S2Zn | 293.57 g/mol
$63.19 - $188.89
Chemical Identifiers
| CAS | 77998-33-5 |
|---|---|
| Molecular Formula | H8N2O8S2Zn |
| Molecular Weight (g/mol) | 293.57 |
| MDL Number | MFCD18711942 |
| InChI Key | CHORECZHRAUKSW-UHFFFAOYSA-L |
| Synonym | ammonium zinc sulfate hydrate, acmc-20alsa, ammonium hydrate zinc sulfate, cvgyqralnwebmp-uhfffaoysa-m, ammonium ? 1-zinc 1+ hydrate sulfate, sulfuric acid, ammoniumzinc salt 2:2:1 , hydrate 9ci |
| PubChem CID | 53471899 |
| SMILES | [NH4+].[NH4+].[Zn++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1358430
|
Thermo Scientific Chemicals
A1358430 |
250 g |
Each for $63.19
|
|
|||||
|
AAA135840B
|
Thermo Scientific Chemicals
A135840B |
1000 g |
Each for $188.89
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 77998-33-5 | |
| 293.57 | |
| CHORECZHRAUKSW-UHFFFAOYSA-L | |
| 53471899 |
| H8N2O8S2Zn | |
| MFCD18711942 | |
| ammonium zinc sulfate hydrate, acmc-20alsa, ammonium hydrate zinc sulfate, cvgyqralnwebmp-uhfffaoysa-m, ammonium ? 1-zinc 1+ hydrate sulfate, sulfuric acid, ammoniumzinc salt 2:2:1 , hydrate 9ci | |
| [NH4+].[NH4+].[Zn++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O |
Specifications
| 77998-33-5 | |
| H8N2O8S2Zn | |
| UN3077 | |
| CHORECZHRAUKSW-UHFFFAOYSA-L | |
| azanium;zinc;sulfate;hydrate | |
| 53471899 | |
| 1.931 |
| 250 g | |
| MFCD18711942 | |
| ammonium zinc sulfate hydrate, acmc-20alsa, ammonium hydrate zinc sulfate, cvgyqralnwebmp-uhfffaoysa-m, ammonium ? 1-zinc 1+ hydrate sulfate, sulfuric acid, ammoniumzinc salt 2:2:1 , hydrate 9ci | |
| [NH4+].[NH4+].[Zn++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O | |
| 293.57 | |
| 293.57 (Anhydrous) | |
| Ammonium zinc sulfate hydrate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only