missing translation for 'onlineSavingsMsg'
Learn More

Ammonium thiosulfate, 96%

Catalog No. p-7080606
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1019022 100 g
AAA1019036 500 g
AAA101900E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1019022 Supplier Thermo Scientific Chemicals Supplier No. A1019022

CAS: 7783-18-8 | (NH4)2S2O3

Ammonium thiosulfate finds application for leaching of gold and silver since it forms strong complexes. It is widely utilized as a photographic fixing agent, which acts quicklier than sodium thiosulfate fixers. It serves as a fertilizer. It acts as an additive to coal-waste mixtures to inhibit the formation of dangerous dioxins and furans. It is used as an analytical reagent in laboratories.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ammonium thiosulfate finds application for leaching of gold and silver since it forms strong complexes. It is widely utilized as a photographic fixing agent, which acts quicklier than sodium thiosulfate fixers. It serves as a fertilizer. It acts as an additive to coal-waste mixtures to inhibit the formation of dangerous dioxins and furans. It is used as an analytical reagent in laboratories.

Solubility
Soluble in water, slightly soluble in acetone. Insoluble in ethanol and diethyl ether.

Notes
Hygroscopic. Incompatible with strong oxidizing agents, acids, magnesium and aluminum.

Chemical Identifiers

CAS 7783-18-8
Molecular Formula (NH4)2S2O3
MDL Number MFCD00015965
Synonym ammonium thiosulphate, thiosulfuric acid h2s2o3 , diammonium salt, thiosulfuric acid h2s2o3 , ammonium salt 1:2, diammonium hypo, acmc-1bhnz, ammoniumthiosulfate, dsstox_cid_9703, dsstox_rid_78814, dsstox_gsid_29703, diammonium thiosulphate

Specifications

Melting Point ∼180°C (decomposition)
pH 6.0 to 6.5
Quantity 100 g
Merck Index 14,562
Solubility Information Soluble in water,slightly soluble in acetone. Insoluble in ethanol and diethyl ether.
Formula Weight 148.2
Percent Purity ≥96%
Sensitivity Hygroscopic
Density 1.64
Chemical Name or Material Ammonium thiosulfate
EINECSNumber 231-982-0
RTECSNumber XN6465000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only