missing translation for 'onlineSavingsMsg'
Learn More

Ammonium thiosulfate, 96%, pure

Catalog No. ac391660025
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
10 kg
2.5 kg
500 g
Packaging:
Plastic bottle
Plastic drum
Catalog No. Quantity Packaging
AC391660025 2.5 kg Plastic bottle
AC391665000 500 g Plastic bottle
AC391660100 10 kg Plastic drum
3 options

Catalog No. AC391660025

Supplier: Thermo Scientific Chemicals 391660025

Only null left

CAS: 7783-18-8 | H8N2O3S2 | 148.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent

Chemical Identifiers

CAS 7783-18-8
Molecular Formula H8N2O3S2
Molecular Weight (g/mol) 148.21
MDL Number MFCD00015965
Synonym ammonium thiosulphate, thiosulfuric acid h2s2o3 , diammonium salt, thiosulfuric acid h2s2o3 , ammonium salt 1:2, diammonium hypo, acmc-1bhnz, ammoniumthiosulfate, dsstox_cid_9703, dsstox_rid_78814, dsstox_gsid_29703, diammonium thiosulphate

Specifications

Melting Point 150.0°C
Color White
Physical Form Crystalline Powder
Quantity 2.5 kg
Assay Percent Range 95% min. (Iodimetry)
Linear Formula (NH4)2S2O3
Merck Index 15, 559
Solubility Information Solubility in water: 63.1% (20°C) - 71.0% (70°C). Other solubilities: insoluble in alcohol and ether
Formula Weight 148.21
Percent Purity 96%
Grade Pure
Packaging Plastic bottle
Chemical Name or Material Ammonium thiosulfate, Extra Pure
Show More Show Less
EINECSNumber 231-982-

RUO – Research Use Only