missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Thiocyanate 98.0+%, TCI America™
Supplier: TCI America A308825G
Specifications
| Ammonium Thiocyanate | |
| 150°C | |
| 4.0 - 5.5 | |
| MFCD00011428 | |
| ammonium thiocyanate, ammoniumrhodanid, thiocyanic acid, ammonium salt, ammonium isothiocyanate, ammonium rhodanate, weedazol tl, trans-aid, ammonium sulfocyanate, ammonium sulfocyanide, rhodanine, ammonium salt | |
| C(#N)[S-].[NH4+] | |
| 76.117 | |
| 76.12 | |
| Crystalline Powder |
| 1762-95-4 | |
| White | |
| CH4N2S | |
| 25 g | |
| SOIFLUNRINLCBN-UHFFFAOYSA-N | |
| azanium;thiocyanate | |
| 15666 | |
| ≥98.0% (T) |
Chemical Identifiers
| 1762-95-4 | |
| 76.117 | |
| SOIFLUNRINLCBN-UHFFFAOYSA-N | |
| 15666 | |
| C(#N)[S-].[NH4+] |
| CH4N2S | |
| MFCD00011428 | |
| ammonium thiocyanate, ammoniumrhodanid, thiocyanic acid, ammonium salt, ammonium isothiocyanate, ammonium rhodanate, weedazol tl, trans-aid, ammonium sulfocyanate, ammonium sulfocyanide, rhodanine, ammonium salt | |
| azanium;thiocyanate |
Safety and Handling
EINECSNumber : (1)-0142
RTECSNumber : XK7875000
TSCA : Yes