missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Thiocyanate, Crystal, Reagent, ACS, 97.5%, Spectrum™ Chemical

Catalog No. p-7156928
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600682 25 g Amber Glass Bottle
18600683 125 g Amber Glass Bottle
18600684 500 g Amber Glass Bottle
18600686 2.5 kg Poly Pail
18600685 6 x 500 g Amber Glass Bottle
18600687 4 x 2.5 kg Poly Pail
18600688 12 kg Poly Pail
18600689 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 18600682 Supplier Spectrum Chemical Mfg Cor Supplier No. A127025GM
Only null left

A1270, 1762-95-4, NH4SCN

Spectrum™ Chemical Ammonium Thiocyanate, Crystal, Reagent, ACS is an inorganic laboratory reagent commonly used by the soft drinks industry to determine the iron content in soft drinks by colorimetry. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 1762-95-4
Molecular Formula CH4N2S
Molecular Weight (g/mol) 76.12
InChI Key SOIFLUNRINLCBN-UHFFFAOYSA-N
IUPAC Name ammonium cyanosulfanide
SMILES [NH4+].[S-]C#N

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 25 g
Percent Purity 97.5%
Grade ACS