missing translation for 'onlineSavingsMsg'
Learn More

Ammonium thiocyanate concentrate, For 1L standard solution, 0.1 M NH4SCN (0.1N), Honeywell Fluka™

Catalog No. 60046678
Change view
Click to view available options
Quantity:
1 Pc.
6 x 1 Ea.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60046678 6 x 1 Ea.
6002655 1 Pc.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60046678 Supplier Honeywell Chemicals Supplier No. 380206X1EA
Only null left

for 1L standard solution, 0.1 M NH4SCN (0.1N)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 1762-95-4 , 7732-18-5
Molecular Formula CH4N2S
Molecular Weight (g/mol) 76.117
MDL Number MFCD00011428
InChI Key SOIFLUNRINLCBN-UHFFFAOYSA-N
Synonym ammonium thiocyanate, ammoniumrhodanid, thiocyanic acid, ammonium salt, ammonium isothiocyanate, ammonium rhodanate, weedazol tl, trans-aid, ammonium sulfocyanate, ammonium sulfocyanide, rhodanine, ammonium salt
PubChem CID 15666
IUPAC Name azanium;thiocyanate
SMILES C(#N)[S-].[NH4+]

Specifications

Boiling Point 100°C
CAS Min % ≥80.0000%, ≥10.0000%
CAS Max % <90.0000%, <20.0000%
Physical Form Concentrate
Quantity 6 x 1 Ea.
Linear Formula NH4SCN
UN Number UN3082
Beilstein 3595135
Formula Weight 76.12
Packaging Ampoule
Chemical Name or Material Ammonium thiocyanate concentrate, For 1L standard solution, 0.1 M NH4SCN (0.1N)
Show More Show Less
ShelfLife 1260 days from date of manufacture
DOTInformation 9 / PGIII
Recommended Storage Room Temp